Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl octafluoroadipate ≥98%

Supplier: MATRIX SCIENTIFIC MS

004847-25G 004847-100G
101787-896EA 272.31 USD InStock
101787-896 101787-898
Dimethyl octafluoroadipate ≥98%
Dimethyl octafluoroadipate
Hazard: Irritant


Formula: C₈H₆F₈O₄
Boiling Pt: 120 °C (15 mmHg)
Density: 1.547
MDL Number: MFCD00042267
CAS Number: 3107-98-0
Order Now
Specification Test Results
Boiling point 120°C/15mm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty