Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Dichloro-3,3,3-trifluoro-2-(trifluoromethyl)propane ≥97%

Supplier: MATRIX SCIENTIFIC MS

005003-1G 005003-5G 005003-25G
101788-140EA 36.92 USD InStock
101788-140 101788-142 101788-144
1,2-Dichloro-3,3,3-trifluoro-2-(trifluoromethyl)propane ≥97%
1,2-Dichloro-3,3,3-trifluoro-2-(trifluoromethyl)propane
Hazard: Irritant


Formula: C₄H₂Cl₂F₆
Boiling Pt: 78-79°C
MDL Number: MFCD01862044
CAS Number: 138690-25-2
Order Now
Specification Test Results
Boiling point 78-79°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty