Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4-Difluorocyclohexylamine hydrochloride ≥97%

Supplier: MATRIX SCIENTIFIC MS

011285-250MG 011285-1G
101800-332EA 84.62 USD InStock
101800-332 101800-334
4,4-Difluorocyclohexylamine hydrochloride ≥97%
4,4-Difluorocyclohexylamine hydrochloride
Hazard: Irritant/Moisture Sensitive


Formula: C₆H₁₂ClF₂N
Boiling Pt: 45…47 °C (12 mmHg)
MDL Number: MFCD06246881
CAS Number: 675112-70-6
Order Now
Specification Test Results
Boiling point 45-47°C/12m

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty