Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α,α-Difluorophenylacetic acid ethyl ester ≥99%

Supplier: MATRIX SCIENTIFIC MS

021136-1G 021136-5G 021136-25G
101811-420EA 38.46 USD InStock
101811-420 101811-422 101811-424
α,α-Difluorophenylacetic acid ethyl ester ≥99%
alpha,alpha-Difluorophenylacetic acid ethyl ester
Hazard: Irritant


Formula: C₁₀H₁₀F₂O₂
MW: 200.19 g/mol
Boiling Pt: 70…72 °C (1 mmHg)
MDL Number: MFCD08275296
CAS Number: 2248-46-6
Order Now
Specification Test Results
Boiling point 70-72°C/1mm

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty