Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2,3-dichloro-5-(trifluoromethyl)isonicotinate ≥95%

Supplier: MATRIX SCIENTIFIC MS

046665-500MG 046665-1G
101876-468EA 921.54 USD InStock
101876-468 101876-470
Methyl-2,3-dichloro-5-(trifluoromethyl)isonicotinate ≥95%
Methyl 2,3-dichloro-5-(trifluoromethyl)isonicotinate
Hazard: Irritant


Formula: C₈H₄NO₂Cl₂F₃
Boiling Pt: 120 °C (0.01 mmHg)
MDL Number: MFCD12025892
CAS Number: 1147979-43-8
Order Now
Specification Test Results
Boiling point 120°C/0.01m

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty