Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2,2-dichloro-2-cyanoacetate ≥95%

Supplier: MATRIX SCIENTIFIC MS

047124-500MG 047124-1G 047124-5G
101878-540EA 461.54 USD InStock
101878-540 101878-542 101878-544
Methyl-2,2-dichloro-2-cyanoacetate ≥95%
Methyl-2,2-dichloro-2-cyanoacetate
Hazard: Irritant


Formula: C₄H₃Cl₂NO₂
Boiling Pt: 67 °C (2 mmHg)
MDL Number: MFCD11100221
CAS Number: 25761-68-6
Order Now
Specification Test Results
Boiling point 67°C/2mm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty