Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-(trifluoromethyl)benzoate ≥97%

Supplier: MATRIX SCIENTIFIC MS

006390-5G 006390-25G
101790-496EA 36.92 USD InStock
101790-496 101790-498
Ethyl-2-(trifluoromethyl)benzoate ≥97%
Ethyl-2-(trifluoromethyl)benzoate
Hazard: Irritant


Formula: C₁₀H₉F₃O₂
Boiling Pt: 90-91°C/10mm
MDL Number: MFCD00013556
CAS Number: 577-62-8
Order Now
Specification Test Results
Boiling point 90-91°C/10m

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty