Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(n-Butoxy)benzenesulfonyl chloride ≥97%

Supplier: MATRIX SCIENTIFIC MS

006542-5G 006542-25G
101790-688EA 73.85 USD InStock
101790-688 101790-690
4-(n-Butoxy)benzenesulfonyl chloride ≥97%
4-(n-Butoxy)benzenesulfonyl chloride
Hazard: Corrosive/Moisture Sensitive


Formula: C₁₀H₁₃ClO₃S
MW: 248.73 g/mol
Boiling Pt: 130…131 °C (0.6 mmHg)
MDL Number: MFCD00052344
CAS Number: 1138-56-3
Order Now
Specification Test Results
Boiling point 130-131°C/0

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty