Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-chlorobenzoylacetate ≥90%

Supplier: MATRIX SCIENTIFIC MS

006946-1G 006946-5G
101791-634EA 98.46 USD InStock
101791-634 101791-636
Methyl-2-chlorobenzoylacetate ≥90%
Methyl-2-chlorobenzoylacetate
Hazard: Irritant


Formula: C₁₀H₉ClO₃
Boiling Pt: 105 °C (0.2 mmHg)
MDL Number: MFCD00216528
CAS Number: 205985-98-4
Order Now
Specification Test Results
Boiling point 105°C/0.2mm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty