Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-(trifluoromethyl)benzoylacetate

Supplier: MATRIX SCIENTIFIC MS

006952-1G 006952-5G
101791-654EA 66.15 USD InStock
101791-654 101791-656
Methyl-2-(trifluoromethyl)benzoylacetate
Methyl-2-(trifluoromethyl)benzoylacetate
Hazard: Irritant


Formula: C₁₁H₉F₃O₃
MW: 246.19 g/mol
Boiling Pt: 98…105 °C (0.5 mmHg)
MDL Number: MFCD00216523
CAS Number: 212755-77-6
Order Now
Specification Test Results
Boiling point 98-105°C/0.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty