Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(-)-1,1,1-Trifluorooctan-2-ol ≥ ee 98%

Supplier: MATRIX SCIENTIFIC MS

007100-1G 007100-5G
101792-020EA 413.85 USD InStock
101792-020 101792-022
(S)-(-)-1,1,1-Trifluorooctan-2-ol ≥ ee 98%
(S)-(-)-1,1,1-Trifluorooctan-2-ol
Hazard: Irritant


Formula: C₈H₁₅F₃O
Boiling Pt: 170°C
Density: 1.05
Flash Pt: 73.3°C
MDL Number: MFCD06799352
CAS Number: 129443-08-9
Order Now
Specification Test Results
Boiling point 75-76°C/20m

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty