Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(-)-1,1,1-Trifluoroundecan-2-ol ≥ ee 98%

Supplier: MATRIX SCIENTIFIC MS

007103-1G 007103-5G
101792-032EA 110.77 USD InStock
101792-032 101792-034
(S)-(-)-1,1,1-Trifluoroundecan-2-ol ≥ ee 98%
(S)-(-)-1,1,1-Trifluoroundecan-2-ol
Hazard: Irritant


Formula: C₁₁H₂₁F₃O
Boiling Pt: 78-79°C/4mm
MDL Number: MFCD06799355
CAS Number: 181933-95-9
Order Now
Specification Test Results
Boiling point 78-79°C/4mm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty