Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Aminophenyl)ethylamine ≥97%

Supplier: MATRIX SCIENTIFIC MS

007453-1G 007453-5G
101792-806EA 60 USD InStock
101792-806 101792-808
2-(4-Aminophenyl)ethylamine ≥97%
2-(4-Aminophenyl)ethylamine
Hazard: Corrosive


Formula: C₈H₁₂N₂
Boiling Pt: 103 °C (0.3 mmHg)
Melting Pt: 28…31 °C
Density: 1.057
MDL Number: MFCD00008194
CAS Number: 13472-00-9
Order Now
Specification Test Results
Boiling point 103°C/0.3mm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty