Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl bromodifluoroacetate ≥98%

Supplier: MATRIX SCIENTIFIC MS

012860-25G 012860-5G
101803-528EA 252.31 USD InStock
101803-528 102832-852
Methyl bromodifluoroacetate ≥98%
Methyl bromodifluoroacetate
Hazard: Flammable/Corrosive/Light Sensitive


Formula: C₃H₃BrF₂O₂
Boiling Pt: 97…98 °C
Density: 1.7125
MDL Number: MFCD06248118
CAS Number: 683-98-7
Order Now
Specification Test Results
Boiling point 97-98°C

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty