Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(2-Chloro-6-fluorophenyl)but-3-en-2-one ≥98%

Supplier: MATRIX SCIENTIFIC MS

024649-5G 024649-25G
101818-548EA 60 USD InStock
101818-548 101818-550
4-(2-Chloro-6-fluorophenyl)but-3-en-2-one ≥98%
4-(2-Chloro-6-fluorophenyl)but-3-en-2-one
Hazard: Irritant


Formula: C₁₀H₈ClFO
Boiling Pt: 115°C/0.8mm
MDL Number: MFCD00051608
CAS Number: 175136-82-0
Order Now
Specification Test Results
Boiling point 115°C/0.8mm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty