Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-1H-1,2,4-triazole ≥98%

Supplier: MATRIX SCIENTIFIC MS

031269-5G 031269-25G
101831-362EA 36.92 USD InStock
101831-362 101831-364
1-Methyl-1H-1,2,4-triazole ≥98%
1-Methyl-1H-1,2,4-triazole
Hazard: Irritant


Formula: C₃H₅N₃
Boiling Pt: 228°C/752 Torr
Melting Pt: 15-16°C
Density: 1.1
Flash Pt: 81°C
MDL Number: MFCD01076192
CAS Number: 6086-21-1
Order Now
Specification Test Results
Boiling point 177-178°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty