Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-methyl-2-(1H-1,2,4-triazol-1-yl)propanoate ≥95%

Supplier: MATRIX SCIENTIFIC MS

042513-500MG 042513-1G 042513-5G
101856-124EA 181.54 USD InStock
101856-124 101856-126 101856-128
Ethyl-2-methyl-2-(1H-1,2,4-triazol-1-yl)propanoate ≥95%
Ethyl-2-methyl-2-(1H-1,2,4-triazol-1-yl)propanoate
Hazard: Irritant


Formula: C₈H₁₃N₃O₂
Boiling Pt: 96 °C (2 mmHg)
MDL Number: MFCD00202105
CAS Number: 100159-13-5
Order Now
Specification Test Results
Boiling point 96°C/2mm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty