Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(3-Fluoropropoxy)benzaldehyde ≥95%

Supplier: MATRIX SCIENTIFIC MS

043032-500MG 043032-1G
101858-806EA 375.38 USD InStock
101858-806 101858-808
4-(3-Fluoropropoxy)benzaldehyde ≥95%
4-(3-Fluoropropoxy)benzaldehyde
Hazard: Irritant


Formula: C₁₀H₁₁FO₂
Boiling Pt: 92-94°C/0.1mm
MDL Number: MFCD02090999
CAS Number: 400825-68-5
Order Now
Specification Test Results
Boiling point 92-94°C/0.1

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty