Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Chloroethyl)-1H-pyrazole ≥95%

Supplier: MATRIX SCIENTIFIC MS

043424-500MG 043424-1G 043424-5G
101860-718EA 35.38 USD InStock
101860-718 101860-720 101860-722
1-(2-Chloroethyl)-1H-pyrazole ≥95%
1-(2-Chloroethyl)-1H-pyrazole
Hazard: Irritant


Formula: C₅H₇ClN₂
Boiling Pt: 60 °C (0.3 mmHg)
MDL Number: MFCD00173067
CAS Number: 96450-53-2
Order Now
Specification Test Results
Boiling point 60°C/0.3mm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty