Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Isobutoxyacetic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

043574-500MG 043574-1G 043574-5G
101861-500EA 227.69 USD InStock
101861-500 101861-502 101861-504
2-Isobutoxyacetic acid ≥95%
2-Isobutoxyacetic acid
Hazard: Irritant/Store in Freezer


Formula: C₆H₁₂O₃
Boiling Pt: 74…76 °C (0.9 mmHg)
MDL Number: MFCD03425725
CAS Number: 24133-46-8
Order Now
Specification Test Results
Boiling point 74-76°C/0.9

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty