Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Formylphenoxy)acetonitrile ≥95%

Supplier: MATRIX SCIENTIFIC MS

045666-500MG 045666-1G 045666-5G
101872-004EA 278.46 USD InStock
101872-004 101872-006 101872-008
2-(4-Formylphenoxy)acetonitrile ≥95%
2-(4-Formylphenoxy)acetonitrile
Hazard: Irritant


Formula: C₉H₇NO₂
MDL Number: MFCD02629373
CAS Number: 385383-45-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty