Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Dimethoxy-1-piperidino-1-ethanone ≥95%

Supplier: MATRIX SCIENTIFIC MS

045791-500MG 045791-1G 045791-5G
101872-618EA 124.62 USD InStock
101872-618 101872-620 101872-622
2,2-Dimethoxy-1-piperidino-1-ethanone ≥95%
2,2-Dimethoxy-1-piperidino-1-ethanone
Hazard: Irritant


Formula: C₉H₁₇NO₃
Boiling Pt: 72 °C (0.01 mmHg)
MDL Number: MFCD08689667
CAS Number: 16695-59-3
Order Now
Specification Test Results
Boiling point 72°C/0.01mm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty