Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-5-fluoro-2-methylphenyl methyl ether ≥95%

Supplier: MATRIX SCIENTIFIC MS

046111-500MG 046111-1G
101874-218EA 649.23 USD InStock
101874-218 101874-220
4-Bromo-5-fluoro-2-methylphenyl methyl ether ≥95%
4-Bromo-5-fluoro-2-methylphenyl methyl ether
Hazard: Irritant


Formula: C₈H₈BrFO
Boiling Pt: 90°C/0.2mm
MDL Number: MFCD11840965
CAS Number: 901231-33-2
Order Now
Specification Test Results
Boiling point 90°C/0.2mm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty