Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2-ethynyl-5-(trifluoromethyl)pyridine ≥95%

Supplier: MATRIX SCIENTIFIC MS

046283-500MG 046283-1G 046283-5G
101874-940EA 556.92 USD InStock
101874-940 101874-942 101874-944
3-Chloro-2-ethynyl-5-(trifluoromethyl)pyridine ≥95%
3-Chloro-2-ethynyl-5-(trifluoromethyl)pyridine
Hazard: Irritant


Formula: C₈H₃ClF₃N
Boiling Pt: 24 °C (0.1 mmHg)
MDL Number: MFCD11840989
CAS Number: 1135283-18-9
Order Now
Specification Test Results
Boiling point 24°C/0.1mm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty