Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-[(Dimethylamino)methyl]-4-(2-methylbutyl)benzenol ≥90%

Supplier: MATRIX SCIENTIFIC MS

046364-500MG 046364-1G 046364-5G
101875-178EA 375.38 USD InStock
101875-178 101875-180 101875-182
2-[(Dimethylamino)methyl]-4-(2-methylbutyl)benzenol ≥90%
2-[(Dimethylamino)methyl]-4-(2-methylbutyl)benzenol
Hazard: Irritant


Formula: C₁₄H₂₃NO
MW: 221.35 g/mol
Boiling Pt: 108 °C (0.01 mmHg)
MDL Number: MFCD11100202
CAS Number: 1033463-17-0
Order Now
Specification Test Results
Boiling point 108°C/0.01m

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty