Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Chloro-4-fluorophenoxy)acetonitrile ≥95%

Supplier: MATRIX SCIENTIFIC MS

046379-500MG 046379-1G 046379-5G
101875-242EA 278.46 USD InStock
101875-242 101875-244 101875-246
2-(2-Chloro-4-fluorophenoxy)acetonitrile ≥95%
2-(2-Chloro-4-fluorophenoxy)acetonitrile
Hazard: Irritant


Formula: C₈H₅ClFNO
MW: 185.59 g/mol
Boiling Pt: 138 °C (12 mmHg)
MDL Number: MFCD09152771
CAS Number: 24115-19-3
Order Now
Specification Test Results
Boiling point 138°C/12mm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty