Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-1-cyanocyclopentanecarboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

046444-500MG 046444-1G 046444-5G
101875-522EA 429.23 USD InStock
101875-522 101875-524 101875-526
Methyl-1-cyanocyclopentanecarboxylate ≥95%
Methyl-1-cyanocyclopentanecarboxylate
Hazard: Irritant


Formula: C₈H₁₁NO₂
Boiling Pt: 80 °C (0.7 mmHg)
MDL Number: MFCD09972231
CAS Number: 40862-12-2
Order Now
Specification Test Results
Boiling point 80°C@0.7mm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty