Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl-2-(dimethylamino)malonate ≥90%

Supplier: MATRIX SCIENTIFIC MS

046454-500MG 046454-1G 046454-5G
101875-560EA 181.54 USD InStock
101875-560 101875-562 101875-564
Diethyl-2-(dimethylamino)malonate ≥90%
Diethyl-2-(dimethylamino)malonate
Hazard: Irritant


Formula: C₉H₁₇NO₄
MW: 203.24 g/mol
Boiling Pt: 46…58 °C (0.1 mmHg)
MDL Number: MFCD00110675
CAS Number: 89222-12-8
Order Now
Specification Test Results
Boiling point 46-58°C/0.1

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty