Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-6-chloropyrazine ≥98%

Supplier: MATRIX SCIENTIFIC MS

001650-25G 001650-100G
101782-184EA 50.77 USD InStock
101782-184 101782-186
2-Amino-6-chloropyrazine ≥98%
2-Amino-6-chloropyrazine
TCA listed: T
Hazard: Irritant


Formula: C₄H₄ClN₃
MW: 129.55 g/mol
Melting Pt: 152…154 °C
MDL Number: MFCD00055024
CAS Number: 33332-28-4
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty