Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2,6-difluorobenzamide

Supplier: MATRIX SCIENTIFIC MS

001965-1G 001965-5G
101782-954EA 30.77 USD InStock
101782-954 101782-956
3-Chloro-2,6-difluorobenzamide
3-Chloro-2,6-difluorobenzamide
Hazard: Irritant


Formula: C₇H₄ClF₂NO
MDL Number: MFCD01631386
CAS Number: 261762-41-8
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty