Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2,6-difluorophenylacetonitrile

Supplier: MATRIX SCIENTIFIC MS

001974-1G 001974-5G
101782-972EA 87.69 USD InStock
101782-972 101782-974
3-Chloro-2,6-difluorophenylacetonitrile
3-Chloro-2,6-difluorophenylacetonitrile
Hazard: Toxic


Formula: C₈H₄ClF₂N
MDL Number: MFCD01631327
CAS Number: 261762-55-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty