Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-5-(trifluoromethyl)cinnamic acid

Supplier: MATRIX SCIENTIFIC MS

002644-1G 002644-5G
101784-378EA 43.08 USD InStock
101784-378 101784-380
3-Fluoro-5-(trifluoromethyl)cinnamic acid
3-Fluoro-5-(trifluoromethyl)cinnamic acid
Hazard: Irritant


Formula: C₁₀H₆F₄O₂
MDL Number: MFCD00236296
CAS Number: 575469-96-4
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty