Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(3-Piperidinyl)ethyl cyclopentanecarboxylate hydrochloride

Supplier: MATRIX SCIENTIFIC MS

049343-500MG 049343-2.5G
101887-276EA 364.62 USD InStock
101887-276 101887-278
2-(3-Piperidinyl)ethyl cyclopentanecarboxylate hydrochloride
2-(3-Piperidinyl)ethyl cyclopentanecarboxylate hydrochloride
Hazard: Irritant


Formula: C₁₃H₂₄ClNO₂
MDL Number: MFCD13559834
CAS Number: 1219980-89-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty