Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Pyrrolidinylmethyl benzoate hydrochloride

Supplier: MATRIX SCIENTIFIC MS

049366-500MG 049366-2.5G
101887-368EA 364.62 USD InStock
101887-368 101887-370
3-Pyrrolidinylmethyl benzoate hydrochloride
3-Pyrrolidinylmethyl benzoate hydrochloride
Hazard: Irritant


Formula: C₁₂H₁₆ClNO₂
MW: 241.72 g/mol
MDL Number: MFCD13559848
CAS Number: 1220027-52-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty