Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-[(3-Chloro[1,1'-biphenyl]-4-yl)oxy]azetidine

Supplier: MATRIX SCIENTIFIC MS

049416-500MG 049416-2.5G
101887-568EA 364.62 USD InStock
101887-568 101887-570
3-[(3-Chloro[1,1'-biphenyl]-4-yl)oxy]azetidine
3-[(3-Chloro[1,1'-biphenyl]-4-yl)oxy]azetidine
Hazard: Irritant


Formula: C₁₅H₁₄ClNO
MDL Number: MFCD13559885
CAS Number: 1219981-44-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty