Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-2-(1H-pyrazol-1-yl)benzaldehyde

Supplier: MATRIX SCIENTIFIC MS

053709-500MG 053709-2.5G
101904-706EA 166.15 USD InStock
101904-706 101904-708
3-Fluoro-2-(1H-pyrazol-1-yl)benzaldehyde
3-Fluoro-2-(1H-pyrazol-1-yl)benzaldehyde
Hazard: Irritant


Formula: C₁₀H₇FN₂O
MDL Number: MFCD09054724
CAS Number: 1214622-47-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty