Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Piperazin-1-yl(pyridin-3-yl)methanone HCl

Supplier: MATRIX SCIENTIFIC MS

053756-500MG 053756-2.5G
101904-890EA 538.46 USD InStock
101904-890 101904-892
Piperazin-1-yl(pyridin-3-yl)methanone HCl
Piperazin-1-yl(pyridin-3-yl)methanone
Hazard: Irritant


Formula: C₁₀H₁₃N₃O
MDL Number: MFCD08456890
CAS Number: 39640-08-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty