Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methyl-1-(2-nitrophenyl)-1H-pyrazole-3-carboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

058681-500MG 058681-1G 058681-5G
101915-382EA 506.15 USD InStock
101915-382 101915-384 101915-386
5-Methyl-1-(2-nitrophenyl)-1H-pyrazole-3-carboxylic acid ≥95%
5-Methyl-1-(2-nitrophenyl)-1H-pyrazole-3-carboxylic acid
Hazard: Irritant


Formula: C₁₁H₉N₃O₄
MW: 247.21 g/mol
MDL Number: MFCD13190657
CAS Number: 103856-74-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty