Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Nitrooxy)butyl butanoate ≥95%

Supplier: MATRIX SCIENTIFIC MS

058689-500MG 058689-1G 058689-5G
101915-420EA 401.54 USD InStock
101915-420 101915-422 101915-424
4-(Nitrooxy)butyl butanoate ≥95%
4-(Nitrooxy)butyl butanoate
Hazard: Irritant


Formula: C₈H₁₅NO₅
MDL Number: MFCD18157662
CAS Number: 1100273-14-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty