Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N'-[2-(4-Chlorophenyl)-1-[(furan-2-ylmethyl)amino]ethylidene]hydrazinecarboxylic acid tert-butyl ester ≥95%

Supplier: MATRIX SCIENTIFIC MS

058892-500MG 058892-1G
101916-216EA 284.62 USD InStock
101916-216 101916-218
N'-[2-(4-Chlorophenyl)-1-[(furan-2-ylmethyl)amino]ethylidene]hydrazinecarboxylic acid tert-butyl ester ≥95%
N'-[2-(4-Chlorophenyl)-1-[(furan-2-ylmethyl)amino]ethylidene]hydrazinecarboxylic acid tert-butyl ester
Hazard: Irritant


Formula: C₁₈H₂₂ClN₃O₃
MDL Number: MFCD10568259
CAS Number: 1053657-85-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty