Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[(Cyano-phenyl-methyl)-amino]-acetic acid ethylester hydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

058943-1G 058943-5G 058943-10G
101916-420EA 320 USD InStock
101916-420 101916-422 101916-424
[(Cyano-phenyl-methyl)-amino]-acetic acid ethylester hydrochloride ≥95%
[(Cyano-phenyl-methyl)-amino]-acetic acid ethylester hydrochloride
Hazard: Irritant


Formula: C₁₂H₁₅ClN₂O₂
MDL Number: MFCD11052443
CAS Number: 854705-52-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty