Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(3-Methoxyphenyl)acetamidoxime hydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

059118-1G 059118-5G 059118-10G
101917-158EA 263.08 USD InStock
101917-158 101917-160 101917-162
2-(3-Methoxyphenyl)acetamidoxime hydrochloride ≥95%
2-(3-Methoxyphenyl)acetamidoxime hydrochloride
Hazard: Irritant


Formula: C₉H₁₃ClN₂O₂
MW: 216.67 g/mol
MDL Number: MFCD10568309
CAS Number: 1208092-47-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty