Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N'-[2-(4-Methoxyphenyl)-1-(2,2,2-trifluoroethylamino)ethylidene]hydrazinecarboxylic acid-tert-butyl ≥95%

Supplier: MATRIX SCIENTIFIC MS

059121-500MG 059121-1G
101917-174EA 284.62 USD InStock
101917-174 101917-176
N'-[2-(4-Methoxyphenyl)-1-(2,2,2-trifluoroethylamino)ethylidene]hydrazinecarboxylic acid-tert-butyl ≥95%
N'-[2-(4-Methoxyphenyl)-1-(2,2,2-trifluoroethylamino)ethylidene]hydrazinecarboxylic acid-tert-butyl
Hazard: Irritant


Formula: C₁₆H₂₂F₃N₃O₃
MDL Number: MFCD10568255
CAS Number: 1053657-61-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty