Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Oxazol-5-ylbenzenesulfonyl chloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

059203-500MG 059203-1G 059203-5G
101917-474EA 363.08 USD InStock
101917-474 101917-476 101917-478
3-Oxazol-5-ylbenzenesulfonyl chloride ≥95%
3-Oxazol-5-ylbenzenesulfonyl chloride
Hazard: Irritant


Formula: C₉H₆ClNO₃S
MW: 243.67 g/mol
MDL Number: MFCD03407346
CAS Number: 499770-75-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty