Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Piperazine-1-carboxamidinium acetate ≥95%

Supplier: MATRIX SCIENTIFIC MS

059228-1G 059228-5G 059228-10G
101917-574EA 320 USD InStock
101917-574 101917-576 101917-578
Piperazine-1-carboxamidinium acetate ≥95%
Piperazine-1-carboxamidinium acetate
Hazard: Irritant


Formula: C₇H₁₆N₄O₂
MDL Number: MFCD10568300
CAS Number: 60729-88-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty