Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Thiophene-3-carboxamidoxime ≥95%

Supplier: MATRIX SCIENTIFIC MS

059251-1G 059251-5G 059251-10G
101917-662EA 407.69 USD InStock
101917-662 101917-664 101917-666
Thiophene-3-carboxamidoxime ≥95%
Thiophene-3-carboxamidoxime
Hazard: Irritant


Formula: C₅H₆N₂OS
MDL Number: MFCD01313358
CAS Number: 58905-71-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty