Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-(1H-pyrrol-1-yl)benzoate

Supplier: MATRIX SCIENTIFIC MS

063296-500MG 063296-2.5G
101925-496EA 476.92 USD InStock
101925-496 101925-498
Methyl-3-(1H-pyrrol-1-yl)benzoate
Methyl-3-(1H-pyrrol-1-yl)benzoate
Hazard: Irritant


Formula: C₁₂H₁₁NO₂
MDL Number: MFCD04082979
CAS Number: 168618-25-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty