Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bromodifluoroacetamide ≥98%

Supplier: MATRIX SCIENTIFIC MS

008829-1G 008829-5G
101795-550EA 47.69 USD InStock
101795-550 101795-552
Bromodifluoroacetamide ≥98%
Bromodifluoroacetamide
Hazard: Irritant


Formula: C₂H₂BrF₂NO
MW: 173.95 g/mol
Boiling Pt: 110…111 °C
Melting Pt: 86 °C
MDL Number: MFCD00193065
CAS Number: 2169-67-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty