Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1-Bromo)perfluoro-2,5,8-trioxanonane ≥98%

Supplier: MATRIX SCIENTIFIC MS

008849-5G 008849-25G
101795-604EA 146.15 USD InStock
101795-604 101795-606
(1-Bromo)perfluoro-2,5,8-trioxanonane ≥98%
(1-Bromo)perfluoro-2,5,8-trioxanonane
Hazard: Irritant


Formula: C₆BrF₁₃O₃
MDL Number: MFCD03094295
CAS Number: 330562-45-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty