Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Difluoroacetamide ≥97%

Supplier: MATRIX SCIENTIFIC MS

008914-5G 008914-25G
101795-808EA 43.08 USD InStock
101795-808 101795-810
2,2-Difluoroacetamide ≥97%
2,2-Difluoroacetamide
Hazard: Irritant


Formula: C₂H₃F₂NO
Boiling Pt: 109°C/35 Torr
Melting Pt: 52 °C
MDL Number: MFCD01896431
CAS Number: 359-38-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty